Medium Pallets, 3 Skids
Lieferant: MULTIROIR
RMP.1210.A.3
MULCRMP.1210.A.3EA 174 EUR
MULCRMP.1210.A.3
Medium Pallets, 3 Skids
Boxen Transportboxen
Medium pallet with safety edge in recycled polypropylene material resistant to shocks and temperature variations.
  • Made of polypropylene for high impact resistance
  • 3 skids
  • Static load 4000 kg
  • Dynamic load 1500 kg
  • High load capacity and light weight
  • Resistant to thermal and weather shocks
  • Colour: Black
Suitable for export to all destinations.