Ross-Miles apparatus
Lieferant: NEUBERT VOLUME GLASSWAERE
2749-90-010
NEUB2749-90-010EA 1850 EUR
NEUB2749-90-010
Ross-Miles apparatus
Apparate zur Mineralöluntersuchung
This Ross-Miles apparatus is used for testing of surfactants.
It is specially necessary during the determination of foaming powder.
Standards: DIN 53902