Magnetic roll holder, PIG®
Lieferant: NEW PIG
57701
NEWP57701EA 13.9 EUR
NEWP57701
Magnetic roll holder, PIG®
Absorptionsmaterial
Magnetic roll holder helps make accessing and storing PIG® Absorbent mats easy and convenient.
  • Holds one 38 cm × 15 m PIG® mat roll for easy one-hand dispensing
  • Durable, powder-coated steel resists chips, rust and corrosion
  • Powerful magnets keep roll holder firmly in place during dispensing; can be removed and relocated in seconds
  • Ideal for attaching to tool boxes, car lifts, support beams or other metal surfaces
Magnetic holder attaches to any metal surface to keep your PIG® Mat roll right where you need it for quick cleanups.