Drum dolly, adjustable
Lieferant: NEW PIG
DLY301-01
NEWPDLY301-01EA 258 EUR
NEWPDLY301-01
Drum dolly, adjustable
Fasslager
This drum dolly with adjustable diameter quickly expands for a precise fit on drums or overpacks up to Ø 677 mm and 363 kg.
  • Heavy-duty 5 mm steel frame adjusts in minutes with bolts and lock nuts
  • Curved ends hold drum tight
  • Wide, stable wheelbase helps prevent tipovers
  • Hard rubber casters for smooth rolling
  • All four casters have brakes to offer secure positioning and keep dolly from moving
Wheel Ø 70 mm