Burpless® One-hand sealable drum funnel, PIG®
Lieferant: NEW PIG
DRM1125-RD-NPT
NEWPDRM1125-RD-NPTEA 369.5 EUR
NEWPDRM1125-RD-NPT
Burpless® One-hand sealable drum funnel, PIG®
Trichter Filtertrichter Fasstrichter
Patented Burpless® designs deliver no-splash filling and one-handed access for easy regulatory compliance.
  • Fast pouring, no splashback, and a one-hand latching lid make it easy to meet regulations
  • Exclusive latching handle design easily opens and closes lid in a simple, single-handed operation
  • Secure closure helps meet closed container regulations, but opens and closes instantly
  • Patented Burpless® design vents during pouring for fast, smooth flow with no dangerous splashback
  • Allows pouring without venting through ³/₄" bung, a possible closed container regulation violation if left open
  • Lockable to prevent unauthorised access
Nitrile gasket and locking mechanism completely seal lid to help reduce vapour emissions. Durable, chemical-resistant powder coat helps ensure long service life.
Use with 115 and 210 litre Steel Drums with 2" BSP bung.
Funnel: 18-gauge steel
Lid gasket: 70 Dura nitrile '87'
Bung gasket: PTFE
Standards: FM approved when used with flame arrestor.