Sani-Trax™ disinfectant mat
Lieferant: NEW PIG
FLME175
NEWPFLME175EA 58.2 EUR
NEWPFLME175
Sani-Trax™ disinfectant mat
Fußmatten Permanente Bodenmatten
Clean and sanitise shoes and prevent contamination from entering controlled areas with this disinfecting footbath.
  • Thousands of flexible fingers dislodge contaminants from footwear using automatic scrubbing action
  • As traffic passes, rubber tips bend under pressure to immerse soles in disinfectant solution, keeping contamination from reaching food processing areas
  • Thick surrounding border stands 6.5mm higher than finger-scrapers to contain up to 4,5 litres of disinfectant solution
  • Convenient mat size that fits most doorways features suction cups on underside to prevent mat from shifting
  • Made from a tough natural rubber compound designed to resist to most chemicals and variations in temperature
Use at doorways, packaging areas, laboratories, locker rooms, break rooms, refuse areas and receiving rooms - anyplace where spread of contamination must be limited.
Bitte beachten Use quaternary ammonium, iodine, chlorine or any other sanitiser but always test its concentration. Most sanitisers are concentrated and must be diluted before use in a footbath. Follow label directions for appropriate dilutions.