PIG® Spill Kits in High-Visibility Containers
Lieferant: NEW PIG
KIT279-OR
NEWPKIT279-OREA 1166.75 EUR
NEWPKIT279-OR
PIG® Spill Kits in High-Visibility Containers
Absorptionsmaterial
These absorbent spill kits are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulphuric acid and 30% sodium hydroxide.
  • UV-resistant polyethylene container keeps absorbents clean and dry indoors or out
  • Highly-visible colour helps to locate the kit faster
  • Absorbency: Up to 287 L
Lieferumfang: PIG Universal includes: 10× blue socks (80×300 mm), 31× blue socks (80×1220 mm), 154× mat pads (380×510 mm), 112× PR40 wipes (300×330 mm), 15× PE disposal bags (910×1520 mm), 6× tamperproof labels, 1× storage chest (1220×790×970 mm) with 130 mm rubber wheels and instruction manual.