Human recombinant p62/SQSTM1 (from E. coli)
Lieferant: Novus Biologicals
NBP1-44490
NOVUNBP1-44490EA 518 EUR
NOVUNBP1-44490
Human recombinant p62/SQSTM1 (from E. coli)
Proteine und Peptide
The Recombinant Human p62/SQSTM1 Protein from Novus Biologicals is derived from E. coli. The Recombinant Human p62/SQSTM1 Protein has been validated for the following applications: SDS-Page.