High Precision Tweezer, 325-SA
Lieferant: OUTILS RUBIS
325-SA
OUTI325-SAEA 25.3 EUR
OUTI325-SA
High Precision Tweezer, 325-SA
Pinzetten
High precision tweezers, antimagnetic/anti-acid SS - serrated handles.
Product height: 1,5 H
Net weight: 11 g