Wafer tweezers, perforated
Lieferant: OUTILS RUBIS
39S-4
OUTI39S-4EA 55.8 EUR
OUTI39S-4
Wafer tweezers, perforated
Pinzetten
Stainless steel. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers.
  • Perforated lower paddle allows liquids to drop off
  • Smooth polished non glare surface on the inside of the paddel in order to prevent chipping and marring the wafer
  • Satin finish on the outside of the paddle for glare prevention