Agar
Lieferant: THERMO FISHER DIAGNOSTICS
Synonyme:
Agar Agar
LP0011T
OXOILP0011TEA
2480
EUR
OXOILP0011T
Agar
Agar
Schmelzpunkt:
90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081288 CAS-Nummer: 9002-18-0 EINECS: 232-658-1 Merck Index: 13,00184 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...