Malzextrakt

Lieferant: THERMO FISHER DIAGNOSTICS

LP0039B
OXOILP0039BEA 81 EUR
OXOILP0039B
Malzextrakt
Malzextrakt
Lagertemperatur: Raumtemperatur
MDL: MFCD00131570
CAS-Nummer: 8002-48-0
EINECS: 232-310-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR