G418 Disulfat, Molekularbiologische Qualität
Lieferant: OZ BIOSCIENCES
|
Gefahr
|
Synonyme:
G-418 Sulfat, G418 Sulfat, Geneticin®, G-418 Disulfat
OZ Biosciences G418 sulphate is an aminoglycoside antibiotic identical to gentamicin B1 produced by Micromonospora rhodorangea.
It blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells. It is used to select and maintain eukaryotic cells expressing the neo gene (neomycin). The quality and purity of the G-418 is essential to achieve good and consistent selection.
Lieferumfang: Shipped at room temperature
Verpackung: Store at -20°C
Formel:
C₂₀H₄₄N₄O₁₈S₂ Molecular Weight: 692,72 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00058314 CAS-Nummer: 108321-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...