D(-)-Luciferin Natriumsalz Monohydrate ≥99,5% endotoxinfrei

Lieferant: OZ BIOSCIENCES

Synonyme: Sodium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate, D-Luciferin, sodium, D-Luciferin sodium, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt

LN10000
OZBILN10000EA 538.65 EUR
OZBILN10000
D(-)-Luciferin Natriumsalz Monohydrate ≥99,5% endotoxinfrei
D(-)-Luciferin sodium salt

OZ Biosciences D-Luciferin K+ and Na+ salts are routinely used as Firefly’s Luciferase substrate in in vitro and in vivo bioluminescent assays. The quality and purity of the D-Luciferin are essential to obtain good and reproducible results. OZ Biosciences is offering high quality of Endotoxin-Free D-Luciferin K+ and Na+ salts.
Store at -20°C and protect from light.


  • High purity > 99.5%
  • Good solubility and great sensitivity
  • Reliable in vivo reporter for bioluminescent assays
  • Endotoxin free (ideal for in vivo application)
  • Quick and easy diffusion throughout the animal


Suitable for bioluminescent assays, whole animal imaging, ATP assays, immunoassays, pyrosequencing, luciferase fragment complementation for sequential gene analysis experiments.

Formel: C₁₁H₇N₂NaO₃S₂
Molecular Weight: 302,30 g/mol
MDL: MFCD00044938
CAS-Nummer: 103404-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af2d873233a094b3a909f57573bdf04fbaec5d4c&vid=812c7d41e42b1b7583f75d25f413b61aa5f82a5a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR