Magnetic separation rack
Lieferant: OZ BIOSCIENCES
MSR1000
OZBIMSR1000EA 340.78 EUR
OZBIMSR1000
Magnetic separation rack
Gestelle Trenngestelle, magnetisch
Magnetic separation rack designed for 50, 15 or 1,5 ml tubes. It can hold 12 standard microtubes, two 15 ml* and two 50 ml tubes. The magnetic separation rack is required for capture, concentration, washing and elution when using Mag4C kits.
  • Can be easily cleaned and decontaminated with 70% ethanol
  • Can be used at room temperature, 37°C, +4°C, etc.
  • Compatible with culture tubes from most common suppliers
  • Solid, completely reusable
*Ø17 mm