di-Natriumhydrogenphosphat Dodecahydrat, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: Natriumphosphat dibasisch Dodecahydrat, sek. Natriumphosphat Dodecahydrat

P103-43EA 0 EUR
P103-43
di-Natriumhydrogenphosphat Dodecahydrat, Macron Fine Chemicals™
di-Natriumhydrogenphosphat Dodecahydrat
Formel: Na₂HPO₄·12H₂O
Molecular Weight: 358,14 g/mol
MDL: MFCD00149181
CAS-Nummer: 10039-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR