di-Natriumhydrogenphosphat Dodecahydrat, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
Natriumphosphat dibasisch Dodecahydrat, sek. Natriumphosphat Dodecahydrat
P110-42EA
464.62
EUR
P110-42
di-Natriumhydrogenphosphat Dodecahydrat, Macron Fine Chemicals™
di-Natriumhydrogenphosphat Dodecahydrat
Formel:
Na₂HPO₄·12H₂O Molecular Weight: 358,14 g/mol |
MDL:
MFCD00149181 CAS-Nummer: 10039-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...