Natriumcarbonat Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: wasserfreie Soda

P120-33EA 0 EUR
P120-33
Natriumcarbonat Ph. Eur., Macron Fine Chemicals™
Natriumcarbonat
Formel: Na₂CO₃
Molecular Weight: 105,99 g/mol
MDL: MFCD00003494
CAS-Nummer: 497-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR