Avantor® Partisil C8, HPLC Columns
Lieferant: VWR
Avantor® Partisil
P5C8-100AEA 942 EUR
P5C8-100A P5C8-250A P5C8-10C5-SK P5C8-10C5 P10C8-150AS P5C8-150A P5C8-250AM P10C8-250P HICH96255/N P10C8-100AM P5C8-100P P10C8-150AM P5C8-150AS P5C8-100AM P10C8-50AM P10C8-250A P10C8-250AM P5C8-150AM HICHP5C8-75A P5C8-100AF P5C8-50AM
Avantor® Partisil C8, HPLC Columns
Chromatographiesäulen
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® C8 is recommended for ion-pair chromatography.
Bitte beachten * Use with freestanding holder HI-161 and column coupler HI-081, please see “HPLC Guard Column Holders and Accessories”.