Ponceau S-Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
P7170-1L
P7170-1LEA
180
EUR
P7170-1L
Ponceau S-Lösung, Sigma-Aldrich®
Ponceau S
Ponceau S solution has been used for membrane staining (Western blotting).
Formel:
C₂₂H₁₂N₄Na₄O₁₃S₄ Molecular Weight: 760,56 g/mol Dichte: 1,01 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003892 CAS-Nummer: 6226-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...