Automatic viscometer
Lieferant: Anton Paar
105001
PAAR105001EA 69300 EUR
PAAR105001
Automatic viscometer
Viskosimeter Automatische Kapillarviskosimetersysteme
Automatic viscometer can measure samples with one integrated cell. SVM 3001 features a wide temperature range. The Peltier temperature control enables fast heating and cooling rates of up to 20 °C/min and the embedded PC provides a wide range of automatic calculations.
  • Multiple parameter measurement
  • Low sample and solvent volume
Automatic VI method, API calculations, temperature scans, FillingCheck™ – Optional countercooling, automation, modularity with Abbemat refractometers.
Standards: EN 61326-1; EN 61010-1; RoHS compliance.