Hastelloy C-276 Polarimeter Tube
Lieferant: Anton Paar
109920
PAAR109920EA 0 EUR
PAAR109920
Hastelloy C-276 Polarimeter Tube
Polarimeter
Hastelloy sample cell (polarimeter tube) 100 mm with wireless Toolmaster function which transfers temperature and path length to the MCP 150.
Filling volume approximately 2 ml. Connection to a syringe through Luer filling ports. Suitable for use with Anton Paar polarimeter.