Tablet reagents for Chlorine Combined and Total (Extended Range DPD XF and XT)
Lieferant: Palintest
PM033
PALIPM033EA
92.8
EUR
PALIPM033
Tablet reagents for Chlorine Combined and Total (Extended Range DPD XF and XT)
Umwelt-Prüfkits
Tablet reagents for chlorine – free, combined and total (extended range DPD XF and XT).
Bestellinformation: Starter Packs (PM codes) contain 50 tests plus any optional reagents or equipment needed.