AcroPrep™ 24-Well Receiver Plate

Lieferant: Cytiva (Formerly Pall Lab)
97028
PALL97028EA 215 EUR
PALL97028
AcroPrep™ 24-Well Receiver Plate
Mikrotiterplatten Einsetz-Platten und Einsätze
Width: 168 mm
Depth: 168 mm
Weight: 70 mm

  • Temperature: Room temperature
  • No shelf life
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR