Pellet presses
Lieferant: PARR
2814
PARR2814EA 0 EUR
PARR2814
Pellet presses
Pressen
Pellet presses used for compressing powdered samples into pellet or tablet form for oxygen bomb calorimetry.
  • Pellets produced in cylindrical shape with flat ends
Suitable for pressing powdered material into briquette form.