Screw top waste and wash vial for autosampler
Lieferant: PerkinElmer
09923031
PERC09923031EA 4.2 EUR
PERC09923031
Screw top waste and wash vial for autosampler
Vials
This screw top waste and wash vial is manufactured from type I borosilicate glass.
Standards: Meets USP, JP, and EP Pharmacopeia requirements.