Reversed phase chromatography cartridge columns, Spheri-5®
Lieferant: PerkinElmer
7110017
PERK7110017EA 0 EUR
PERK7110017
Reversed phase chromatography cartridge columns, Spheri-5®
Chromatographiesäulen
Spheri-5® is a small pore silica based sorbent for separating small molecules.