HA-Tag Magnetic IP/Co-IP Kit, Pierce™
Lieferant: Thermo Fisher Scientific
88838
PIER88838EA 804 EUR
PIER88838
HA-Tag Magnetic IP/Co-IP Kit, Pierce™
Protein-Aufbereitungs Protein Purification Kits
The Pierce™ HA-Tag Magnetic IP/Co-IP Kit contains specific immunoaffinity magnetic beads and reagents to perform immunoprecipitation assays of HA fusion proteins or co-IP experiments using HA-tagged bait proteins.
  • Covalently immobilised high-quality anti-HA monoclonal antibody enables high yields of immunoprecipitation products
  • Stable, pre-blocked beads and specific antibody minimise off-target binding
  • Complete kit and easy to follow instructions provide optimised protocols to perform IP or Co-IP experiments in approximately 1 hour
  • Magnetic beads are compatible with manual and automated magnetic separation workflows
Lieferumfang: The kit includes HA-tagged positive control , 500 µl; anti-HA magnetic beads, 1 ml; IP lysis/wash buffer, 2×50 ml; non-reducing sample buffer (5X), 5 ml; elution buffer (pH 2,0), 5 ml; neutralization buffer (pH 8,5), 1 ml.