Magnetic separation rack, MagJET

Lieferant: Thermo Fisher Scientific
MR02
PIERMR02EA 460 EUR
PIERMR02
Magnetic separation rack, MagJET
Gestelle Trenngestelle, magnetisch
This separation rack is a magnetic separation device intended for low-throughput magnetic bead manipulation procedures such as magnetic bead-based nucleic acid isolation.

  • Contains a powerful permanent neodymium magnet for instantaneous bead collection from the solution
  • Specially designed for robust performance with different sample volumes
  • Stable and tight magnetic bead capture on microcentrifuge tube wall
  • Highly efficient nucleic acid purification in combination with Thermo Scientific MagJET magnetic-bead based kits

The MagJET separation rack contains a high-performance permanent neodymium magnet embedded in a plastic separation unit frame. In combination with MagJET magnetic beads the separation aack allows fast and efficient magnetic bead-based separation procedures.

Applications: Affinity purification using magnetic particles.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR