Glutaraldehyd 8% in wässriger Lösung
Lieferant: POLYSCIENCES
|
Gefahr
|
Synonyme:
Glutaral, Pentandial, 1,5-Pentandial
00216-30
POLS00216-30EA
0
EUR
POLS00216-30
Glutaraldehyd 8% in wässriger Lösung
Glutaraldehyd
Formel:
C₅H₈O₂ Molecular Weight: 100,12 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00007025 CAS-Nummer: 111-30-8 EINECS: 203-856-5 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...