Storage bottles, narrow neck, round, PTFE, with screw caps
Lieferant: Poulten & Graf
10.1305-54
POUL10.1305-54EA 424 EUR
POUL10.1305-54
Storage bottles, narrow neck, round, PTFE, with screw caps
Flaschen Kunststoffflaschen
PTFE, white.
  • Excellent chemical resistance
  • Heat resistant from –200 up to +280 °C
  • Exceptionally smooth inner walls to prevent contamination
Robust, unbreakable construction. The special recessed grips on the cap and grooves in the flask make it easy to handle. Non stick, the wide mouth allows easy filling and cleaning.