Boric acid isotopic standard, NIST-951A
Lieferant: LGC Standards PROMOCHEM
NIST-951A
PROONIST-951AEA
767
EUR
PROONIST-951A
Boric acid isotopic standard, NIST-951A
Standards
Umweltanalyse-Standards
This standard reference material is intended for use as an isotopic reference material for the calibration of mass spectrometers. The material consists of highly purified boric acid of high homogeneity.
Mixture of approx 20% Boron 10 and 80% Boron 11.
Standards: Traceable to NIST and SRM.