Earmuffs, helmet-mounted, Endurance, PW47
Lieferant: Portwest
PW47RER
PRTWPW47REREA 15.9 EUR
PRTWPW47RER
Earmuffs, helmet-mounted, Endurance, PW47
Kapselgehörschützer Lärmschutz-Kapselgehörschützer
These helmet-mounted ear protectors made from ABS feature an excellent attenuation level.
  • Can be adjusted into stand-off position when not in use
  • Easy to clip on
Compatible with Portwest hemlets PS53, PW54 and PW55.
SNR: 29 dB
Standards: EN 352-3