PYROTELL®: Gel-clot formulation (Method A and B, EP)
25003A-05 25006A-05 20125A 20003A 20006A 20250A 20003A-50 20006A-50 20125A-50 20250A-50 25003A-25
PYRQ25003A-05EA 0 EUR
PYRQ25003A-05 PYRQ25006A-05 PYRQ20125A PYRQ20003A PYRQ20006A PYRQ20250A PYRQ20003A-50 PYRQ20006A-50 PYRQ20125A-50 PYRQ20250A-50 PYRQ25003A-25
PYROTELL®: Gel-clot formulation (Method A and B, EP)
Mikrobiologische Testsysteme Endotoxin-Tests
Pyrotell® lysate was the first LAL gel-clot reagent licensed by the US FDA. It is easy to use and is available in both economical multi-test vials and convenient Single Test Vials (STV’s). Pyrotell lysate is a robust reagent, producing firm, easily read clots and is resistant to interfering substances. The gel-clot test does not require sophisticated capital equipment and software and is the simplest LAL test to implement.