PYROTELL®: Gel-clot formulation (Method A and B, EP)
25003A-05
25006A-05
20125A
20003A
20006A
20250A
20003A-50
20006A-50
20125A-50
20250A-50
25003A-25
PYRQ25003A-05EA
0
EUR
PYRQ25003A-05
PYRQ25006A-05
PYRQ20125A
PYRQ20003A
PYRQ20006A
PYRQ20250A
PYRQ20003A-50
PYRQ20006A-50
PYRQ20125A-50
PYRQ20250A-50
PYRQ25003A-25
PYROTELL®: Gel-clot formulation (Method A and B, EP)
Mikrobiologische Testsysteme
Endotoxin-Tests
Pyrotell® lysate was the first LAL gel-clot reagent licensed by the US FDA. It is easy to use and is available in both economical multi-test vials and convenient Single Test Vials (STV’s). Pyrotell lysate is a robust reagent, producing firm, easily read clots and is resistant to interfering substances. The gel-clot test does not require sophisticated capital equipment and software and is the simplest LAL test to implement.