Xylencyanol FF

Lieferant: G Bioscienes

Synonyme: C.I. 42135, Sodium hydrogen 4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulphonate, Sodium 4-((4-(ethylamino)-3-methylphenyl)(4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-sulphobenzenesulphonate, acid Blue 147

RC-115
RC-115EA 303 EUR
RC-115
Xylencyanol FF
Xylene Cyanole FF

A colour marker to monitor the process of agarose gel electrophoresis and polyacrylamide gel electrophoresis.


In 1% agarose gels, Xylene cyanol typically migrates at about the same rate as a 4000 base pair DNA fragment. Xylene cyanol or tracking dye on a 6% polacrylamide gel migrates at the speed of a 140 base pair DNA fragment.

Formel: C₂₅H₂₇N₂NaO₆S₂
Molecular Weight: 538,62 g/mol
Schmelzpunkt: 295 °C
MDL: MFCD00040657
CAS-Nummer: 2650-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR