Disposable Specimen Cuvettes for UNISTAT Bilirubinometers
Lieferant: REICHERT
13102140
REIC13102140EA 143 EUR
REIC13102140
Disposable Specimen Cuvettes for UNISTAT Bilirubinometers
Küvetten Spezialküvetten
Disposable specimen cuvettes specifically designed for UNISTAT Bilirubinometers. These cuvettes hold 20 μl (microliters) of serum and are placed into the optical path of our Bilirubinometer to measure newborn Total Serum Bilirubin concentration.
Verpackung: 51 disposable cuvettes are supplied in a package.