Gas regulators
Lieferant: Restek
21666
REST21666EA 593 EUR
REST21666
Gas regulators
Druckminderer
Chromium plated brass line regulator.
  • Oxidation resistant
  • Same purity protection as high pressure cylinder regulators