Inland 45 pump oil
Lieferant: Restek
24819
REST24819EA 117 EUR
REST24819
Inland 45 pump oil
Schmiermittel
Inland 45 pump oil is recommended for most mass spectrometers. It has low vapor pressure,10⁻⁷ torr and optimum vacuum pump performance.
  • Non-toxic and non-corrosive
  • Compatible with buna-N, neoprene, and Viton® seals