Mobile phase degasser
Lieferant: Restek
25194
REST25194EA 0 EUR
REST25194
Mobile phase degasser
Entgaser
Mobile phase degasser used to remove the dissolved gas from mobile phases. This is better than the sonication or helium sparging techniques.
  • Withdraws gas across a gas-permeable membrane encased in a sealed chamber
  • Degasser tubings are made up of Amorphous Fluoropolymer (AF)
  • Provides stable flow rate and low baseline noise
  • Prevents from quenching effect on fluorescence detection and increased background of UV detectors