DNA (Deoxyribonucleic acid) (aus Kalbsthymus)

Lieferant: Rockland Immunochemicals

MB-102-0100
ROCKMB-102-0100EA 179 EUR
ROCKMB-102-0100
DNA (Deoxyribonucleic acid) (aus Kalbsthymus)
DNS (Desoxyribonukleinsäure)

DNA from calf thymus is commonly used in the preparation of pre-hybridisation and hybridisation solutions. Ideal for DNA polymerase assays. These prepared solutions are used to block the non-specific attachment of probe to the surface of a membrane. The DNA solutions are prepared from highly pure, phenol/chloroform-extracted DNA, and DNase- and RNase-free (DEPC-treated) distilled, deionised water.

MDL: MFCD00130922
CAS-Nummer: 9007-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR