SynaptoRed™ C2 ≥95% (durch HPLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: FM® 4-64, Pyridinium, 4-[6-[4-(diethylamino)phenyl]- 1,3,5-hexatrienyl]-1-[3-(triethylammonio) propyl]-, dibromide, SynaptoRed™, MM 4-64

M-Clarity™ Qualitätsstufe = MQ100

S6689-5MG
S6689-5MGEA 622 EUR
S6689-5MG
SynaptoRed™ C2 ≥95% (durch HPLC), Sigma-Aldrich®
N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64)

SynaptoRed™ C2 has been used for propidium iodide (PI) blockage.


SynaptoRed is a trademark of Biotium, Inc.

Formel: C₃₀H₄₅Br₂N₃
Molecular Weight: 607,51 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00467819
CAS-Nummer: 162112-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5f607e205f75be41d9c649f7c5d8854700803b1&vid=c6918b211c8cd508d81a5efacba9a34f19e190b0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR