X-ray and metal detectable scrapers
Lieferant: Sampling Systems
M1019B-110
SAMPM1019B-110EA 42.1 EUR
SAMPM1019B-110
X-ray and metal detectable scrapers
Schaber
The X-ray and metal detectable scraper is ideal for scraping and mixing material. The scrapers are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the material in line with HACCP guidelines.
  • Robust PP construction, provides excellent chemical resistance to many products
  • Handy blade, simple to use
  • Full batch traceability
To ensure the cleanliness of the scrapers are manufactured and packed in a cleanroom. They can be used as a single use disposable product straight from the bag or reused.
Standards: These scrapers conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Verpackung: Supplied individually bagged.