Aspirating pipettes
Lieferant: SARSTEDT
86.1252.011
SARS86.1252.011EA 227 EUR
SARS86.1252.011
Aspirating pipettes
Pipetten Aspirationspipetten
PS pipettes, non graduated, unplugged, sterile.
  • For aspirating media with a vacuum pump
  • Certified non pyrogenic
  • Individually wrapped in paper plastic peel packaging