This syringe filter holder is made from PTFE. It is well suited for particle removal from samples and reagents for analytical methods, such as NMR samples as it is unaffected by chemicals and contains no trace elements which could be released into the liquid being filtered.
Low hold-up volume
Easy cleaning and drying at a temperature of 180 °C
Leak proof sealing without the use of sealing ring