Nessler colour comparison tubes, KIMBLE®
Lieferant: DWK Life Sciences
SP45310-50100
SCERSP45310-50100EA 417 EUR
SCERSP45310-50100
Nessler colour comparison tubes, KIMBLE®
Röhrchen Nessler-Rohre
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Shadowless bottom of each tube transmits undistorted light
  • Rings and legend are durable white ceramic enamel
  • Matched or unmatched available
These Nessler tubes are suitable for colour comparison.
Standards: Glass conforms to USP Type I and ASTM E438 Type I Class A requirements.