Homogeniser disposable pestles and tubes, polypropene, KIMBLE®
Lieferant: DWK Life Sciences
SP749540-0000
SCERSP749540-0000EA 502 EUR
SCERSP749540-0000
Homogeniser disposable pestles and tubes, polypropene, KIMBLE®
Homogenisatoren Tragbare Homogenisatoren
Stainless steel cordless drive unit. Pressure is 14.7 PSI.
  • Resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
  • Operates at 2000 to 3000 min⁻¹ with fresh batteries and no load
Lieferumfang: Supplied with 2× AA batteries.