BioDrop µLite+ Microvolume Spectrophotometer
Lieferant: BIOCHROM
80-3006-55
SCLI80-3006-55EA 12300 EUR
SCLI80-3006-55
BioDrop µLite+ Microvolume Spectrophotometer
Spektrophotometer und Photometer Biophotometer
BioDrop's unique sample port is dedicated to microvolume measurement. The port is easy to use: simply pipette as little as 0,5 μl and measure. Cleaning the port is easy too, just wipe with lint-free tissue to reduce sample carryover to undetectable amounts.
  • The in-built sample port uses no moving parts
  • Instrument provides excellent reproducibility without the need to recondition or calibrate
  • Highly accurate with the pathlength of the port specified to ±5 μm
  • Export data via USB or link to PC
Easy to use onboard analysis with pre-programmed applications or users designed protocols.
Bestellinformation: BioDrop Resolution CFR software is available if needed for full 12 CFR Part 11 compliance. IQ/OQ available upon request.