Ring light, VisiLED Slim
Lieferant: SCHOTT AG Lighting and imaging
400150
SCOC400150EA 826 EUR
SCOC400150
Ring light, VisiLED Slim
Beleuchtung Ringlichter
The VisiLED Slim is a space saving LED ring light, optimal for unconfined object preparation or usage with objective revolvers.
  • Easily adaptable to microscope objectives by means of thumb screw
  • Homogeneous incident or oblique illumination
  • Controllable with all VisiLED controllers (MC750, MC1000, MC1500)
  • Variable working distance ranges possible by exchange of focus optic rings
  • Implementation of brightfield and darkfield applications
  • Segments controllable in different preset modes