Segmented polariser set for VisiLED ring light, S80-55
Lieferant: SCHOTT AG Lighting and imaging
400555
SCOC400555EA 662 EUR
SCOC400555
Segmented polariser set for VisiLED ring light, S80-55
Beleuchtung Ringlichter
The segmented polariser set has the ability to switch between polarised and non-polarised light, without any mechanical movements. The set is only compatible to VisiLED ringlight S80 55, together with the MC 1500 or the MC 1100 controller.
  • Very helpful for all applications that need polarised and non-polarised light, such as cleanliness inspections
  • Software control possible (MC 1500)
  • Intensities independently adjustable for polarisation and brightfield (MC 1500)
Designed for use with VisiLED ring light S80-55, together with the MC 1500 or the MC 1100 controller.
Bestellinformation: VisiLED ringlight S80-55 and controller MC 1100 or MC 1500 must be purchased seperately.