Gas washing bottles, DURAN®
Lieferant: DWK Life Sciences
257040101
SCOT257040101EA 101 EUR
SCOT257040101
Gas washing bottles, DURAN®
Flaschen Gaswaschflaschen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw cap system with Drechsel-head with filter disc, adjustable immersion.